missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Cyanophenyl)benzimidazole-6-carboxylic acid, 97%

Manufacturer:  Alfa Aesar™ H5424006

 View more versions of this product

Catalog No. AAH5424006

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-(4-Cyanophenyl)benzimidazole-6-carboxylic acid
175401-95-3
MFCD20265213
HRUUSKALOILBKF-UHFFFAOYSA-N
2-(4-cyanophenyl)-3H-benzimidazole-5-carboxylic acid
23509758
97%
5g
C15H9N3O2
1h-benzimidazole-6-carboxylicacid, 2-4-cyanophenyl
C1=CC(=CC=C1C#N)C2=NC3=C(N2)C=C(C=C3)C(=O)O
263.256
263.26

Chemical Identifiers

175401-95-3
263.256
HRUUSKALOILBKF-UHFFFAOYSA-N
23509758
C1=CC(=CC=C1C#N)C2=NC3=C(N2)C=C(C=C3)C(=O)O
C15H9N3O2
MFCD20265213
1h-benzimidazole-6-carboxylicacid, 2-4-cyanophenyl
2-(4-cyanophenyl)-3H-benzimidazole-5-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Documents