missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Cyanophenyl)benzimidazole-6-carboxylic acid, 97%

Manufacturer:  Alfa Aesar™ H5424003

 View more versions of this product

Catalog No. AAH5424003

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-(4-Cyanophenyl)benzimidazole-6-carboxylic acid
175401-95-3
MFCD20265213
HRUUSKALOILBKF-UHFFFAOYSA-N
2-(4-cyanophenyl)-3H-benzimidazole-5-carboxylic acid
23509758
97%
1g
C15H9N3O2
1h-benzimidazole-6-carboxylicacid, 2-4-cyanophenyl
C1=CC(=CC=C1C#N)C2=NC3=C(N2)C=C(C=C3)C(=O)O
263.256
263.26

Chemical Identifiers

175401-95-3
263.256
HRUUSKALOILBKF-UHFFFAOYSA-N
23509758
C1=CC(=CC=C1C#N)C2=NC3=C(N2)C=C(C=C3)C(=O)O
C15H9N3O2
MFCD20265213
1h-benzimidazole-6-carboxylicacid, 2-4-cyanophenyl
2-(4-cyanophenyl)-3H-benzimidazole-5-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents