Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Chlorophenylthio)nicotinic acid, 98%

$19.91 - $114.81

Chemical Identifiers

Chemical Name or Material 2-(4-Chlorophenylthio)nicotinic acid
CAS 955-54-4
Molecular Formula C12H8ClNO2S
Formula Weight 265.72g/mol
MDL Number MFCD00052117
Synonym 2-(4-Chlorophenylthio)pyridine-3-carboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1466103

 
alfa aesar™
A14661-03
1g Each for $19.91

AAA1466106

 
alfa aesar™
A14661-06
5g Each for $114.81
Description & Specifications

Specifications

Melting Point 219° to 221°C
Quantity 1g
Chemical Name or Material 2-(4-Chlorophenylthio)nicotinic acid
CAS 955-54-4
Assay 98%
Molecular Formula C12H8ClNO2S
Formula Weight 265.72g/mol
MDL Number MFCD00052117
Synonym 2-(4-Chlorophenylthio)pyridine-3-carboxylic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.