Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Chlorophenylthio)-6-fluorobenzonitrile, 97%

$44.01 - $483.34

Chemical Identifiers

Chemical Name or Material 2-(4-Chlorophenylthio)-6-fluorobenzonitrile
CAS 175204-12-3
Molecular Formula C13H7ClFNS
Formula Weight 263.72
MDL Number MFCD00068208
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2523003

 
alfa aesar™
B2523003
1g Each for $44.01

AAB2523006

 
alfa aesar™
B2523006
5g Each for $153.93

AAB2523014

 
alfa aesar™
B2523014
25g Each for $483.34
Description & Specifications

Specifications

Formula Weight 263.72
Chemical Name or Material 2-(4-Chlorophenylthio)-6-fluorobenzonitrile
Assay 97%
Molecular Formula C13H7ClFNS
MDL Number MFCD00068208
CAS 175204-12-3
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 68° to 71°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.