Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Chlorophenylsulfonyl)thioacetamide, 97%

$52.96 - $367.89

Chemical Identifiers

Chemical Name or Material 2-(4-Chlorophenylsulfonyl)thioacetamide
CAS 59865-87-1
Molecular Formula C8H8ClNO2S2
Formula Weight 249.73
MDL Number MFCD00052824
Synonym 2-(4-Chlorobenzenesulfonyl)thioacetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2680603

 
alfa aesar™
H2680603
1g Each for $123.00

AAH2680606

 
alfa aesar™
H2680606
5g Each for $367.89

AAH26806MD

 
alfa aesar™
H26806MD
250mg Each for $52.96
Description & Specifications

Specifications

Formula Weight 249.73
Chemical Name or Material 2-(4-Chlorophenylsulfonyl)thioacetamide
Assay 97%
Molecular Formula C8H8ClNO2S2
MDL Number MFCD00052824
CAS 59865-87-1
Synonym 2-(4-Chlorobenzenesulfonyl)thioacetamide
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 186° to 190°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.