Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Chlorophenyl)thioacetamide, 97%

$86.50 - $256.15

Chemical Identifiers

Chemical Name or Material 2-(4-Chlorophenyl)thioacetamide
CAS 17518-48-8
Molecular Formula C8H8ClNS
Formula Weight 185.67
MDL Number MFCD00278249
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2686403

  SDS
alfa aesar™
H2686403
1g Each for $86.50

AAH2686406

  SDS
alfa aesar™
H2686406
5g Each for $256.15
Description & Specifications

Specifications

Formula Weight 185.67
Quantity 1g
Health Hazard 1 Danger
Chemical Name or Material 2-(4-Chlorophenyl)thioacetamide
Melting Point 127° to 130°C
Molecular Formula C8H8ClNS
Assay 97%
MDL Number MFCD00278249
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 17518-48-8

Applications
2-(4-Chlorophenyl)thioacetamide is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.