missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(4-Chlorophenyl)thiazolidine-4-carboxylic acid, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H62341MD
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-(4-Chlorophenyl)thiazolidine-4-carboxylic acid | |
White | |
MFCD00278428,MFCD01413971 | |
2-4-chlorophenyl-1,3-thiazolidine-4-carboxylic acid, 2-4-chlorophenyl-1,3-thiazolane-4-carboxylic acid, 2-4-chlorophenyl thiazolidine-4-carboxylic acid, 2-4-chloro-phenyl-thiazolidine-4-carboxylic acid, maybridge1_000958, 4-thiazolidinecarboxylicacid, 2-4-chlorophenyl, 2-4-chlorophenyl-1,3-thiazolidine-4-carboxylicacid | |
OC(=O)C1CSC(N1)C1=CC=C(Cl)C=C1 | |
243.71 | |
243.71 | |
Solid |
34491-29-7 | |
C10H10ClNO2S | |
250 mg | |
LESQASCTNMKKPZ-UHFFFAOYNA-N | |
2-(4-chlorophenyl)-1,3-thiazolidine-4-carboxylic acid | |
327597 | |
97% |
Chemical Identifiers
34491-29-7 | |
243.71 | |
LESQASCTNMKKPZ-UHFFFAOYNA-N | |
327597 | |
OC(=O)C1CSC(N1)C1=CC=C(Cl)C=C1 |
C10H10ClNO2S | |
MFCD00278428,MFCD01413971 | |
2-4-chlorophenyl-1,3-thiazolidine-4-carboxylic acid, 2-4-chlorophenyl-1,3-thiazolane-4-carboxylic acid, 2-4-chlorophenyl thiazolidine-4-carboxylic acid, 2-4-chloro-phenyl-thiazolidine-4-carboxylic acid, maybridge1_000958, 4-thiazolidinecarboxylicacid, 2-4-chlorophenyl, 2-4-chlorophenyl-1,3-thiazolidine-4-carboxylicacid | |
2-(4-chlorophenyl)-1,3-thiazolidine-4-carboxylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures