missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Chlorophenyl)thiazolidine-4-carboxylic acid, 97%

Manufacturer:  Alfa Aesar™ H6234106

 View more versions of this product

Catalog No. AAH6234106



Specifications

Specifications

2-(4-Chlorophenyl)thiazolidine-4-carboxylic acid
5g
C10H10ClNO2S
2-4-chlorophenyl-1,3-thiazolidine-4-carboxylic acid, 2-4-chlorophenyl-1,3-thiazolane-4-carboxylic acid, 2-4-chlorophenyl thiazolidine-4-carboxylic acid, 2-4-chloro-phenyl-thiazolidine-4-carboxylic acid, maybridge1_000958, 4-thiazolidinecarboxylicacid, 2-4-chlorophenyl, 2-4-chlorophenyl-1,3-thiazolidine-4-carboxylicacid
C1C(NC(S1)C2=CC=C(C=C2)Cl)C(=O)O
243.705
243.71
97%
White
34491-29-7
MFCD00278428
LESQASCTNMKKPZ-UHFFFAOYSA-N
2-(4-chlorophenyl)-1,3-thiazolidine-4-carboxylic acid
327597
Solid

Chemical Identifiers

34491-29-7
243.705
LESQASCTNMKKPZ-UHFFFAOYSA-N
327597
C1C(NC(S1)C2=CC=C(C=C2)Cl)C(=O)O
C10H10ClNO2S
MFCD00278428
2-4-chlorophenyl-1,3-thiazolidine-4-carboxylic acid, 2-4-chlorophenyl-1,3-thiazolane-4-carboxylic acid, 2-4-chlorophenyl thiazolidine-4-carboxylic acid, 2-4-chloro-phenyl-thiazolidine-4-carboxylic acid, maybridge1_000958, 4-thiazolidinecarboxylicacid, 2-4-chlorophenyl, 2-4-chlorophenyl-1,3-thiazolidine-4-carboxylicacid
2-(4-chlorophenyl)-1,3-thiazolidine-4-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents