Please login to your online account to display your discounted pricing

2-[(4-chlorophenyl)sulfonyl]ethanethioamide, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  SPB01724EA

 View more versions of this product

Catalog No. SPB01724EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 249.74
Quantity 10g
Assay 97%
CAS Min % 97.0
Linear Formula ClC6H4SO2CH2CSNH2
Chemical Name or Material 2-[(4-Chlorophenyl)sulfonyl]ethanethioamide
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C8H8ClNO2S2
MDL Number MFCD00052824
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 59865-87-1
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

Specifications

Chemical Name or Material 2-[(4-Chlorophenyl)sulfonyl]ethanethioamide
CAS 59865-87-1
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H8ClNO2S2
Linear Formula ClC6H4SO2CH2CSNH2
Formula Weight 249.74
MDL Number MFCD00052824
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.