missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Chlorophenyl)ethylboronic acid pinacol ester, 98%

$52.49 - $173.93

Chemical Identifiers

CAS 444094-88-6
Molecular Formula C14H20BClO2
Formula Weight 266.58
MDL Number MFCD12546184
Synonym 2-[2-(4-Chlorophenyl)ethyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Chemical Name or Material 2-(4-Chlorophenyl)ethylboronic acid pinacol ester
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3030303 SDS Alfa Aesar™
H3030303
1g Each for $52.49
AAH3030306 SDS Alfa Aesar™
H3030306
5g Each for $173.93
Specifications Description & Specifications

Chemical Identifiers

444094-88-6
266.58
2-[2-(4-Chlorophenyl)ethyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
C14H20BClO2
MFCD12546184
2-(4-Chlorophenyl)ethylboronic acid pinacol ester

Specifications

34° to 38°C
444094-88-6
266.58
2-[2-(4-Chlorophenyl)ethyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
98%
1g
C14H20BClO2
MFCD12546184
2-(4-Chlorophenyl)ethylboronic acid pinacol ester