Please login to your online account to display your discounted pricing

2-(4-Chlorophenyl)ethylamine 98%, ACROS Organics™


Quantity 5mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC109880050

 View more versions of this product

Catalog No. AC109880050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥97.5%
Formula Weight 155.63
Quantity 5mL
Color Clear colorless to yellow
EINECS Number 205-853-4
TSCA TSCA
Density 1.112g/mL
Beilstein 12, IV, 2489
CAS Max % 100.0
Water 0.5% max. (K.F.)
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.5474 to 1.5494 (20°C, 589nm)
Chemical Name or Material 2-(4-Chlorophenyl)ethylamine
Name Note 98%
Assay Percent Range 98%
Molecular Formula C8H10ClN
MDL Number MFCD00008191
CAS 156-41-2
Packaging Glass bottle
Linear Formula ClC6H4CH2CH2NH2
Synonym 4-Chlorophenethylamine
CAS Min % 97.5

Specifications

Chemical Name or Material 2-(4-Chlorophenyl)ethylamine
Name Note 98%
CAS 156-41-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H10ClN
Linear Formula ClC6H4CH2CH2NH2
Formula Weight 155.63
MDL Number MFCD00008191
Synonym 4-Chlorophenethylamine
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents