missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(4-Chlorophenyl)ethylamine 98%, ACROS Organics™

$146.26 - $146.26

Chemical Identifiers

CAS 156-41-2
Molecular Formula C8H10ClN
Molecular Weight (g/mol) 155.625
MDL Number MFCD00008191
InChI Key SRXFXCKTIGELTI-UHFFFAOYSA-N
Synonym 4-chlorophenethylamine, 2-4-chlorophenyl ethylamine, 2-4-chlorophenyl ethanamine, 2-p-chlorophenyl ethylamine, p-chlorophenethylamine, 2-4-chlorophenyl ethan-1-amine, 4-chlorobenzeneethanamine, 2-4-chloro-phenyl-ethylamine, benzeneethanamine, 4-chloro, beta-p-chlorophenyl ethylamine
PubChem CID 67430
IUPAC Name 2-(4-chlorophenyl)ethanamine
SMILES C1=CC(=CC=C1CCN)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC109880050 SDS View Documents Acros Organics
AC109880050
5mL Glass bottle N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC109880250 SDS View Documents Acros Organics
109880250
25mL Glass bottle Each for $146.26
Specifications

Chemical Identifiers

156-41-2
155.625
SRXFXCKTIGELTI-UHFFFAOYSA-N
67430
C1=CC(=CC=C1CCN)Cl
C8H10ClN
MFCD00008191
4-chlorophenethylamine, 2-4-chlorophenyl ethylamine, 2-4-chlorophenyl ethanamine, 2-p-chlorophenyl ethylamine, p-chlorophenethylamine, 2-4-chlorophenyl ethan-1-amine, 4-chlorobenzeneethanamine, 2-4-chloro-phenyl-ethylamine, benzeneethanamine, 4-chloro, beta-p-chlorophenyl ethylamine
2-(4-chlorophenyl)ethanamine

Specifications

1.1120g/mL
Authentic
1.5474 to 1.5494
242.0°C to 245.0°C
5mL
97.5
98%
ClC6H4CH2CH2NH2
12, IV, 2489
SRXFXCKTIGELTI-UHFFFAOYSA-N
2-(4-chlorophenyl)ethanamine
67430
Liquid
2-(4-Chlorophenyl)ethylamine, 98%
106°C
Glass bottle
0.5% max. (K.F.)
Colorless to Yellow
156-41-2
100.0
C8H10ClN
MFCD00008191
4-chlorophenethylamine, 2-4-chlorophenyl ethylamine, 2-4-chlorophenyl ethanamine, 2-p-chlorophenyl ethylamine, p-chlorophenethylamine, 2-4-chlorophenyl ethan-1-amine, 4-chlorobenzeneethanamine, 2-4-chloro-phenyl-ethylamine, benzeneethanamine, 4-chloro, beta-p-chlorophenyl ethylamine
C1=CC(=CC=C1CCN)Cl
155.625
155.63
98%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 205-853-4

TSCA : TSCA