missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(4-Chlorophenyl)ethyl isothiocyanate, 97%, Alfa Aesar™

Manufacturer:  Alfa Aesar L1130706

 View more versions of this product

Catalog No. AAL1130706



Specifications

Specifications

2-(4-Chlorophenyl)ethyl isothiocyanate
Glass bottle
5g
C9H8ClNS
UN2922
MRJJYUJULSZFDV-UHFFFAOYSA-N
1-chloro-4-(2-isothiocyanatoethyl)benzene
140257
97%
>110°C
145°C (1mmHg)
17608-10-5
MFCD00041127
2-4-chlorophenyl ethyl isothiocyanate, 1-chloro-4-2-isothiocyanatoethyl benzene, 2-4-chlorophenethyl isothiocyanate, 2-4-chlorophenethyl isothicyanate, 2-4-chlorophenethyl isothiocyanate, acmc-1bsas, 2-4-chlorophenyl ethanisothiocyanate, 2-4-chlorophenethyl isothiocyanate, 1-chloranyl-4-2-isothiocyanatoethyl benzene, 1-chloro-4-2-isothiocyanatoethyl benzene #
C1=CC(=CC=C1CCN=C=S)Cl
197.68
197.69

Chemical Identifiers

17608-10-5
197.68
MRJJYUJULSZFDV-UHFFFAOYSA-N
140257
C1=CC(=CC=C1CCN=C=S)Cl
C9H8ClNS
MFCD00041127
2-4-chlorophenyl ethyl isothiocyanate, 1-chloro-4-2-isothiocyanatoethyl benzene, 2-4-chlorophenethyl isothiocyanate, 2-4-chlorophenethyl isothicyanate, 2-4-chlorophenethyl isothiocyanate, acmc-1bsas, 2-4-chlorophenyl ethanisothiocyanate, 2-4-chlorophenethyl isothiocyanate, 1-chloranyl-4-2-isothiocyanatoethyl benzene, 1-chloro-4-2-isothiocyanatoethyl benzene #
1-chloro-4-(2-isothiocyanatoethyl)benzene
Safety and Handling

Safety and Handling

TSCA : No

Documents