missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Chlorophenyl)-7-methyl-5-oxo-5H-imidazo[1,2-a]pyrimidine-8-acetic acid

$81.23 - $282.73

Chemical Identifiers

CAS 902836-44-6
Molecular Formula C15H12ClN3O3
Formula Weight 317.73
MDL Number MFCD08064257
Chemical Name or Material 2-(4-Chlorophenyl)-7-methyl-5-oxo-5H-imidazo[1,2-a]pyrimidine-8-acetic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5080803 SDS Alfa Aesar™
H5080803
1g Each for $282.73
AAH50808MD SDS Alfa Aesar™
H50808MD
250mg Each for $81.23
Specifications Description & Specifications

Chemical Identifiers

902836-44-6
317.73
2-(4-Chlorophenyl)-7-methyl-5-oxo-5H-imidazo[1,2-a]pyrimidine-8-acetic acid
C15H12ClN3O3
MFCD08064257

Specifications

1g
C15H12ClN3O3
MFCD08064257
902836-44-6
317.73
2-(4-Chlorophenyl)-7-methyl-5-oxo-5H-imidazo[1,2-a]pyrimidine-8-acetic acid

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning