Please login to your online account to display your discounted pricing

2-(4-Chlorophenyl)-3-oxopropanenitrile, 95%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-(4-Chlorophenyl)-3-oxopropanenitrile
CAS 62538-21-0
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C9H6ClNO
Linear Formula ClC6H4CH(CN)CHO
Formula Weight 179.61
MDL Number MFCD00052669
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SPB01054EA

  SDS
fisher scientific company
SPB01054EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 179.61
Quantity 10g
Assay 95%
CAS Min % 95.0
Assay Percent Range 95%
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2-(4-Chlorophenyl)-3-oxopropanenitrile
Molecular Formula C9H6ClNO
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful in contact with skin.
MDL Number MFCD00052669
Packaging Amber Glass Bottle
Linear Formula ClC6H4CH(CN)CHO
CAS 62538-21-0