Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Chlorophenoxy)thioacetamide, 97%

$219.62 - $219.62

Chemical Identifiers

Chemical Name or Material 2-(4-Chlorophenoxy)thioacetamide
CAS 35368-44-6
Molecular Formula C8H8ClNOS
Formula Weight 201.68g/mol
MDL Number MFCD00052481
Beilstein 1952884
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1271106

 
alfa aesar™
L12711-06
5g Each for $219.62
Description & Specifications

Specifications

Melting Point 104° to 106°C
Quantity 5g
Chemical Name or Material 2-(4-Chlorophenoxy)thioacetamide
CAS 35368-44-6
Assay 97%
Molecular Formula C8H8ClNOS
Formula Weight 201.68g/mol
MDL Number MFCD00052481
Beilstein 1952884