Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Chlorophenoxy)nicotinic acid, 98%

$128.08 - $128.08

Chemical Identifiers

Chemical Name or Material 2-(4-Chlorophenoxy)nicotinic acid
CAS 51362-37-9
Molecular Formula C12H8ClNO3
Formula Weight 249.64
MDL Number MFCD00052116
Synonym 2-(4-Chlorophenoxy)pyridine-3-carboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1435906

 
alfa aesar™
A1435906
5g Each for $128.08
Description & Specifications

Specifications

Formula Weight 249.64
Molecular Formula C12H8ClNO3
MDL Number MFCD00052116
Melting Point 157° to 159°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 2-(4-Chlorophenoxy)nicotinic acid
Health Hazard 1 Warning
Assay 98%
CAS 51362-37-9
Synonym 2-(4-Chlorophenoxy)pyridine-3-carboxylic acid
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.