Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Chlorophenoxy)isobutyric acid, 98%

$33.55 - $84.04

Chemical Identifiers

Chemical Name or Material 2-(4-Chlorophenoxy)isobutyric acid
CAS 882-09-7
Molecular Formula C10H11ClO3
Formula Weight 214.65
MDL Number MFCD00004192
Beilstein 1874067
Merck Index 14,2378
Synonym 2-(4-Chlorophenoxy)-2-methylpropionic acid; Clofibric acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1762414

 
alfa aesar™
A1762414
25g Each for $33.55

AAA1762422

 
alfa aesar™
A1762422
100g Each for $84.04
Description & Specifications

Specifications

Formula Weight 214.65
Chemical Name or Material 2-(4-Chlorophenoxy)isobutyric acid
Assay 98%
Molecular Formula C10H11ClO3
MDL Number MFCD00004192
CAS 882-09-7
Beilstein 1874067
Merck Index 14,2378
Synonym 2-(4-Chlorophenoxy)-2-methylpropionic acid; Clofibric acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number UE9455000
Melting Point 119° to 120°C
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.