Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Chlorophenoxy)ethanol, 98+%

Chemical Identifiers

Chemical Name or Material 2-(4-Chlorophenoxy)ethanol
CAS 1892-43-9
Molecular Formula C8H9ClO2
Formula Weight 172.61
MDL Number MFCD00014036
UN Number UN2811
Beilstein 1365684
Synonym 4-(2-Hydroxyethoxy)chlorobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1450806

  SDS
alfa aesar™
A1450806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1450822

  SDS
alfa aesar™
A1450822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 172.61
Chemical Name or Material 2-(4-Chlorophenoxy)ethanol
TSCA TSCA
Assay ≥98%
Molecular Formula C8H9ClO2
MDL Number MFCD00014036
CAS 1892-43-9
UN Number UN2811
Beilstein 1365684
Synonym 4-(2-Hydroxyethoxy)chlorobenzene
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H318-H312-H335-H315
Causes serious eye damage.
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECS Number KK2100000
Boiling Point 135° to 136°C (6mmHg)
Melting Point 30°C
Quantity 5g

Applications
2-(4-Chlorophenoxy)ethanol is used as a pharmaceutical intermediate.

Solubility
Soluble in water 3.1 g/L at 25°C.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H318-H312-H335-H315
Causes serious eye damage.
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.