missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Chlorophenoxy)ethanol, 98+%

Manufacturer:  Alfa Aesar™ A1450806

 View more versions of this product

Catalog No. AAA1450806



Description

Description

Applications
2-(4-Chlorophenoxy)ethanol is used as a pharmaceutical intermediate.

Solubility
Soluble in water 3.1 g/L at 25°C.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications

Specifications

2-(4-Chlorophenoxy)ethanol
30°C
1892-43-9
MFCD00014036
1365684
GEGSSUSEWOHAFE-UHFFFAOYSA-N
2-(4-chlorophenoxy)ethanol
15907
≥98%
135°C to 136°C (6mmHg)
5g
C8H9ClO2
UN2811
2-4-chlorophenoxy ethanol, chlorophetanol, fungisan, ethanol, 2-4-chlorophenoxy, chloro-p-phenoxetol, 2-p-chlorophenoxy ethanol, p-chlorophenyl glycol ether, p-chlorophenyl monoglycol ether, p-chlorfenylmonoglykolether, ethanol, 2-p-chlorophenoxy
C1=CC(=CC=C1OCCO)Cl
172.608
172.61

Chemical Identifiers

1892-43-9
172.608
GEGSSUSEWOHAFE-UHFFFAOYSA-N
15907
C1=CC(=CC=C1OCCO)Cl
C8H9ClO2
MFCD00014036
2-4-chlorophenoxy ethanol, chlorophetanol, fungisan, ethanol, 2-4-chlorophenoxy, chloro-p-phenoxetol, 2-p-chlorophenoxy ethanol, p-chlorophenyl glycol ether, p-chlorophenyl monoglycol ether, p-chlorfenylmonoglykolether, ethanol, 2-p-chlorophenoxy
2-(4-chlorophenoxy)ethanol
Safety and Handling

Safety and Handling

GHS H Statement
H318-H312-H335-H315
Causes serious eye damage.
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : KK2100000

TSCA : TSCA

Documents