Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Chlorophenoxy)-6-fluoro-N-methylbenzylamine, 99%

$321.73 - $321.73

Chemical Identifiers

Chemical Name or Material 2-(4-Chlorophenoxy)-6-fluoro-N-methylbenzylamine
CAS 902836-83-3
Molecular Formula C14H13ClFNO
Formula Weight 265.72g/mol
MDL Number MFCD08064236
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5073303

 
alfa aesar™
H50733-03
Each for $321.73
Description & Specifications

Specifications

Quantity 1g
Chemical Name or Material 2-(4-Chlorophenoxy)-6-fluoro-N-methylbenzylamine
CAS 902836-83-3
Assay 99%
Molecular Formula C14H13ClFNO
Formula Weight 265.72g/mol
MDL Number MFCD08064236
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.