Please login to your online account to display your discounted pricing

2-(4-chlorophenoxy)-2-methylpropanenitrile, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-(4-chlorophenoxy)-2-methylpropanenitrile
CAS 24889-11-0
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H10ClNO
Linear Formula ClC6H4OC(CH3)2CN
Formula Weight 195.65
MDL Number MFCD00203862
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

RDP00164DA

 
fisher scientific company
RDP00164DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

RDP00164EA

 
fisher scientific company
RDP00164EA
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 195.65
Quantity 1g
Assay 97%
CAS Min % 97.0
Molecular Formula C10H10ClNO
MDL Number MFCD00203862
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Harmful if inhaled.
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
Chemical Name or Material 2-(4-chlorophenoxy)-2-methylpropanenitrile
Assay Percent Range 97%
Linear Formula ClC6H4OC(CH3)2CN
CAS 24889-11-0
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Amber Glass Bottle
Safety and Handling

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Harmful if inhaled.
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.