Please login to your online account to display your discounted pricing

2-[4-(chloromethyl)piperidino]pyrimidine, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC20724DA

 View more versions of this product

Catalog No. CC20724DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 211.69
Quantity 1g
Assay 97%
Chemical Name or Material 2-[4-(chloromethyl)piperidino]pyrimidine
Packaging Amber Glass Bottle
CAS 111247-62-2
Molecular Formula C10H14ClN3

Specifications

Chemical Name or Material 2-[4-(chloromethyl)piperidino]pyrimidine
Assay 97%
Molecular Formula C10H14ClN3
Formula Weight 211.69