Please login to your online account to display your discounted pricing

2-[4-(Chloromethyl)phenyl]-5-(trifluoromethyl)pyridine, 95%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC53024CB

 View more versions of this product

Catalog No. CC53024CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 271.67
Quantity 250mg
Assay 95%
CAS Max % 100.0
Assay Percent Range 95%
CAS 613239-76-2
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 94.5
MDL Number MFCD09817479
Packaging Amber Glass Bottle
Chemical Name or Material 2-[4-(Chloromethyl)phenyl]-5-(trifluoromethyl)pyridine
Molecular Formula C13H9ClF3N

Specifications

Chemical Name or Material 2-[4-(Chloromethyl)phenyl]-5-(trifluoromethyl)pyridine
CAS 613239-76-2
CAS Min % 94.5
CAS Max % 100.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C13H9ClF3N
Formula Weight 271.67
MDL Number MFCD09817479
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents