Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Chlorobenzylthio)ethanol, 98%

$148.85 - $575.69

Chemical Identifiers

Chemical Name or Material 2-(4-Chlorobenzylthio)ethanol
CAS 71501-40-1
Molecular Formula C9H11ClOS
Formula Weight 202.7
MDL Number MFCD00020614
Beilstein 8975850
Synonym 4-Chlorobenzyl 2-hydroxyethyl sulfide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1015806

  SDS
alfa aesar™
L1015806
5g Each for $148.85

AAL1015814

  SDS
alfa aesar™
L1015814
25g Each for $575.69
Description & Specifications

Specifications

Formula Weight 202.7
Chemical Name or Material 2-(4-Chlorobenzylthio)ethanol
Assay 98%
Molecular Formula C9H11ClOS
MDL Number MFCD00020614
CAS 71501-40-1
Beilstein 8975850
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
RTECS Number KK1150000
Boiling Point 139° to 140°C (1mmHg)
Synonym 4-Chlorobenzyl 2-hydroxyethyl sulfide
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.