missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Chlorobenzylthio)ethanol, 98%

Manufacturer:  Alfa Aesar™ L1015814

 View more versions of this product

Catalog No. AAL1015814



Specifications

Specifications

2-(4-Chlorobenzylthio)ethanol
25g
C9H11ClOS
8975850
DKNBCVYGWWWYEP-UHFFFAOYSA-N
2-[(4-chlorophenyl)methylsulfanyl]ethanol
198536
98%
139°C to 140°C (1mmHg)
71501-40-1
MFCD00020614
ethanol, 2-p-chlorobenzylthio, 2-p-chlorobenzylthioethanol, 2-4-chlorobenzylthio ethanol, 2-4-chlorophenyl methyl thio ethanol, 4-chlorobenzyl-2-hydroxyethyl-sulfide, 2-4-chlorophenyl methyl sulfanyl ethanol, 2-4-chlorobenzyl thio ethanol, 2-4-chlorobenzyl sulfanyl ethanol, 2-4-chlorophenyl methylsulfanyl ethanol, 2-4-chlorophenyl methylthio ethan-1-ol
C1=CC(=CC=C1CSCCO)Cl
202.696
202.7

Chemical Identifiers

71501-40-1
202.696
DKNBCVYGWWWYEP-UHFFFAOYSA-N
198536
C1=CC(=CC=C1CSCCO)Cl
C9H11ClOS
MFCD00020614
ethanol, 2-p-chlorobenzylthio, 2-p-chlorobenzylthioethanol, 2-4-chlorobenzylthio ethanol, 2-4-chlorophenyl methyl thio ethanol, 4-chlorobenzyl-2-hydroxyethyl-sulfide, 2-4-chlorophenyl methyl sulfanyl ethanol, 2-4-chlorobenzyl thio ethanol, 2-4-chlorobenzyl sulfanyl ethanol, 2-4-chlorophenyl methylsulfanyl ethanol, 2-4-chlorophenyl methylthio ethan-1-ol
2-[(4-chlorophenyl)methylsulfanyl]ethanol
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

RTECSNumber : KK1150000

Documents