Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Chlorobenzyl)pyridine, 97%

$92.22 - $92.22

Chemical Identifiers

Chemical Name or Material 2-(4-Chlorobenzyl)pyridine
CAS 4350-41-8
Molecular Formula C12H10ClN
Formula Weight 203.67
MDL Number MFCD00006353
Beilstein 133425
Synonym 4-Chlorophenyl 4-pyridyl ketone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2317014

  SDS
alfa aesar™
B2317014
25g Each for $92.22
Description & Specifications

Specifications

Formula Weight 203.67
Assay 97%
Beilstein 133425
Boiling Point 181° to 183°C (20mmHg)
Quantity 25g
Chemical Name or Material 2-(4-Chlorobenzyl)pyridine
Molecular Formula C12H10ClN
MDL Number MFCD00006353
CAS 4350-41-8
Synonym 4-Chlorophenyl 4-pyridyl ketone
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Flash Point >110°C (230°F)
Melting Point 8°C
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.