Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Chlorobenzoyl)benzoic acid, 99%

$22.46 - $186.84

Chemical Identifiers

Chemical Name or Material 2-(4-Chlorobenzoyl)benzoic acid
CAS 85-56-3
Molecular Formula C14H9ClO3
Formula Weight 260.68
MDL Number MFCD00002474
Beilstein 649894
Synonym 2-Carboxy-4'-chlorobenzophenone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0274614

  SDS
alfa aesar™
L0274614
25g Each for $22.46

AAL0274622

  SDS
alfa aesar™
L0274622
100g Each for $52.59

AAL0274636

  SDS
alfa aesar™
L0274636
500g Each for $186.84
Description & Specifications

Specifications

Formula Weight 260.68
Molecular Formula C14H9ClO3
Chemical Name or Material 2-(4-Chlorobenzoyl)benzoic acid
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Synonym 2-Carboxy-4'-chlorobenzophenone
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
CAS 85-56-3
Health Hazard 1 Warning
Beilstein 649894
TSCA TSCA
Assay 99%
MDL Number MFCD00002474
Quantity 25g
Melting Point 149° to 151°C
Odor Odorless
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.