Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Chloro-2-methylphenoxy)acetic acid hydrazide, 96%

$65.24 - $229.03

Chemical Identifiers

Chemical Name or Material 2-(4-Chloro-2-methylphenoxy)acetic acid hydrazide
CAS 32022-38-1
Molecular Formula C9H11ClN2O2
Formula Weight 214.65
MDL Number MFCD00082720
Beilstein 983137
Synonym 2-(4-chloro-o-tolyloxy)acetic acid hydrazide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2452106

  SDS
alfa aesar™
B2452106
5g Each for $65.24

AAB2452114

  SDS
alfa aesar™
B2452114
25g Each for $229.03
Description & Specifications

Specifications

Formula Weight 214.65
Chemical Name or Material 2-(4-Chloro-2-methylphenoxy)acetic acid hydrazide
Assay 96%
Molecular Formula C9H11ClN2O2
MDL Number MFCD00082720
CAS 32022-38-1
Beilstein 983137
Synonym 2-(4-chloro-o-tolyloxy)acetic acid hydrazide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 149° to 151°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.