missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(4-Bromophenyl)thiophene, TCI America™

$157.36 - $465.98

Chemical Identifiers

CAS 40133-22-0
Molecular Formula C10H7BrS
Molecular Weight (g/mol) 239.13
MDL Number MFCD05664340
InChI Key XKOJJKOSNQXQGP-UHFFFAOYSA-N
PubChem CID 7018298
IUPAC Name 2-(4-bromophenyl)thiophene
SMILES C1=CSC(=C1)C2=CC=C(C=C2)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B51061G View Documents TCI America
B51061G
1g Each for $157.36
 
B51065G View Documents TCI America
B51065G
5g Each for $465.98
 
Specifications

Chemical Identifiers

40133-22-0
239.13
XKOJJKOSNQXQGP-UHFFFAOYSA-N
2-(4-bromophenyl)thiophene
C10H7BrS
MFCD05664340
7018298
C1=CSC(=C1)C2=CC=C(C=C2)Br

Specifications

White-Yellow
1g
C10H7BrS
XKOJJKOSNQXQGP-UHFFFAOYSA-N
2-(4-bromophenyl)thiophene
7018298
Crystal-Powder at 20°C
2-(4-Bromophenyl)thiophene
98°C
40133-22-0
MFCD05664340
C1=CSC(=C1)C2=CC=C(C=C2)Br
239.13
239.13
≥97.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No