Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Bromophenyl)imidazo[1,2-a]pyridine

$67.88 - $237.05

Chemical Identifiers

Chemical Name or Material 2-(4-Bromophenyl)imidazo[1,2-a]pyridine
CAS 34658-66-7
Molecular Formula C13H9BrN2
Formula Weight 273.13
MDL Number MFCD00218250
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5059003

  SDS
alfa aesar™
H5059003
1g Each for $237.05

AAH50590MD

  SDS
alfa aesar™
H50590MD
250mg Each for $67.88
Description & Specifications

Specifications

Formula Weight 273.13
Molecular Formula C13H9BrN2
CAS 34658-66-7
Quantity 1g
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 2-(4-Bromophenyl)imidazo[1,2-a]pyridine
MDL Number MFCD00218250
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Applications
2-(4-Bromophenyl)imidazo[1,2-a]pyridine is an important raw material and intermediate used in pharmaceuticals, agrochemicals and dyestuff.

Notes
Incompatible with oxidizing agents. Store in a cool, dry, well-ventilated area.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.