missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(4-Bromophenyl)benzothiazole, TCI America™

$140.56 - $522.52

Chemical Identifiers

CAS 19654-19-4
Molecular Formula C13H8BrNS
Molecular Weight (g/mol) 290.178
InChI Key FQIRBKKYMJKENC-UHFFFAOYSA-N
Synonym 1-(2-Benzothiazolyl)-4-bromobenzene
PubChem CID 2320602
IUPAC Name 2-(4-bromophenyl)-1,3-benzothiazole
SMILES C1=CC=C2C(=C1)N=C(S2)C3=CC=C(C=C3)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B26795G View Documents TCI America
B26795G
5g Each for $140.56
 
B267925G View Documents TCI America
B267925G
25g Each for $522.52
 
Specifications

Chemical Identifiers

19654-19-4
290.178
1-(2-Benzothiazolyl)-4-bromobenzene
2-(4-bromophenyl)-1,3-benzothiazole
C13H8BrNS
FQIRBKKYMJKENC-UHFFFAOYSA-N
2320602
C1=CC=C2C(=C1)N=C(S2)C3=CC=C(C=C3)Br

Specifications

Undesignated
5g
C13H8BrNS
FQIRBKKYMJKENC-UHFFFAOYSA-N
2-(4-bromophenyl)-1,3-benzothiazole
2320602
Crystal-Powder at 20°C
2-(4-Bromophenyl)benzothiazole
134°C
19654-19-4
1-(2-Benzothiazolyl)-4-bromobenzene
C1=CC=C2C(=C1)N=C(S2)C3=CC=C(C=C3)Br
290.178
290.18
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No