missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Bromophenyl)benzimidazole, 95%

$40.55 - $479.59

Chemical Identifiers

CAS 2622-74-4
Molecular Formula C13H9BrN2
Molecular Weight (g/mol) 273.133
MDL Number MFCD00224360
InChI Key YRWMGOSKROWAIT-UHFFFAOYSA-N
Synonym 2-4-bromophenyl benzimidazole, 2-4-bromophenyl-1h-benzo d imidazole, 2-4-bromophenyl-1h-benzimidazole, 2-4-bromophenyl-1h-benzoimidazole, 2-4-bromophenyl benzoimidazole, 2-4-bromo-phenyl-1h-benzoimidazole, 1h-benzimidazole, 2-4-bromophenyl, 2-4-bromophenyl-1h-1,3-benzodiazole, acmc-209gpy, boron-mol bm885
PubChem CID 286182
IUPAC Name 2-(4-bromophenyl)-1H-benzimidazole
SMILES C1=CC=C2C(=C1)NC(=N2)C3=CC=C(C=C3)Br
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64811MD View Documents Alfa Aesar™
H64811MD
250mg Each for $40.55
 
AAH6481103 View Documents Alfa Aesar™
H6481103
1g Each for $120.14
 
AAH6481106 View Documents Alfa Aesar™
H6481106
5g Each for $479.59
 
Specifications

Chemical Identifiers

2622-74-4
273.133
YRWMGOSKROWAIT-UHFFFAOYSA-N
286182
C1=CC=C2C(=C1)NC(=N2)C3=CC=C(C=C3)Br
C13H9BrN2
MFCD00224360
2-4-bromophenyl benzimidazole, 2-4-bromophenyl-1h-benzo d imidazole, 2-4-bromophenyl-1h-benzimidazole, 2-4-bromophenyl-1h-benzoimidazole, 2-4-bromophenyl benzoimidazole, 2-4-bromo-phenyl-1h-benzoimidazole, 1h-benzimidazole, 2-4-bromophenyl, 2-4-bromophenyl-1h-1,3-benzodiazole, acmc-209gpy, boron-mol bm885
2-(4-bromophenyl)-1H-benzimidazole

Specifications

250mg
C13H9BrN2
162670
YRWMGOSKROWAIT-UHFFFAOYSA-N
2-(4-bromophenyl)-1H-benzimidazole
286182
Solid
2-(4-Bromophenyl)benzimidazole
2622-74-4
MFCD00224360
2-4-bromophenyl benzimidazole, 2-4-bromophenyl-1h-benzo d imidazole, 2-4-bromophenyl-1h-benzimidazole, 2-4-bromophenyl-1h-benzoimidazole, 2-4-bromophenyl benzoimidazole, 2-4-bromo-phenyl-1h-benzoimidazole, 1h-benzimidazole, 2-4-bromophenyl, 2-4-bromophenyl-1h-1,3-benzodiazole, acmc-209gpy, boron-mol bm885
C1=CC=C2C(=C1)NC(=N2)C3=CC=C(C=C3)Br
273.133
273.13
95%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning