Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Bromophenyl)acetamidoxime, 97%

Chemical Identifiers

Chemical Name or Material 2-(4-Bromophenyl)acetamidoxime
CAS 422560-40-5
Formula Weight 229.07
MDL Number MFCD05662829
Synonym 2-(4-Bromophenyl)-N-hydroxyacetamidine; 2-(4-Bromophenyl)-1-(hydroxyimino)ethylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6689103

 
alfa aesar™
H6689103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6689106

 
alfa aesar™
H6689106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66891MD

 
alfa aesar™
H66891MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 132° to 134°C
Formula Weight 229.07
Chemical Name or Material 2-(4-Bromophenyl)acetamidoxime
Assay 97%
CAS 422560-40-5
Synonym 2-(4-Bromophenyl)-N-hydroxyacetamidine; 2-(4-Bromophenyl)-1-(hydroxyimino)ethylamine
Physical Form Crystalline Solid
Color White
MDL Number MFCD05662829
Quantity 1g