missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(4-Bromophenyl)-5-phenylthiophene, TCI America™

$122.22 - $366.67

Chemical Identifiers

CAS 118621-30-0
Molecular Formula C16H11BrS
Molecular Weight (g/mol) 315.228
MDL Number MFCD21648452
InChI Key YMDCOEOZWHXGTM-UHFFFAOYSA-N
PubChem CID 10313583
IUPAC Name 2-(4-bromophenyl)-5-phenylthiophene
SMILES C1=CC=C(C=C1)C2=CC=C(S2)C3=CC=C(C=C3)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B5491200MG View Documents TCI America
B5491200MG
200mg Each for $122.22
 
B54911G View Documents TCI America
B54911G
1g Each for $366.67
 
Specifications

Chemical Identifiers

118621-30-0
315.228
YMDCOEOZWHXGTM-UHFFFAOYSA-N
2-(4-bromophenyl)-5-phenylthiophene
C16H11BrS
MFCD21648452
10313583
C1=CC=C(C=C1)C2=CC=C(S2)C3=CC=C(C=C3)Br

Specifications

Undesignated
200mg
C16H11BrS
YMDCOEOZWHXGTM-UHFFFAOYSA-N
2-(4-bromophenyl)-5-phenylthiophene
10313583
Crystal-Powder at 20°C
2-(4-Bromophenyl)-5-phenylthiophene
190°C
118621-30-0
MFCD21648452
C1=CC=C(C=C1)C2=CC=C(S2)C3=CC=C(C=C3)Br
315.228
315.23
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No