Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Bromophenyl)-5-phenyl-1,3,4-oxadiazole, 98+%

$384.82 - $384.82

Chemical Identifiers

Chemical Name or Material 2-(4-Bromophenyl)-5-phenyl-1,3,4-oxadiazole
CAS 21510-43-0
Molecular Formula C14H9BrN2O
Formula Weight 301.14
MDL Number MFCD00050997
Beilstein 204891
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0870106

 
alfa aesar™
L0870106
5g Each for $384.82
Description & Specifications

Specifications

Formula Weight 301.14
Chemical Name or Material 2-(4-Bromophenyl)-5-phenyl-1,3,4-oxadiazole
Assay ≥98%
Molecular Formula C14H9BrN2O
MDL Number MFCD00050997
CAS 21510-43-0
Melting Point 167° to 172°C
Beilstein 204891
Quantity 5g