missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Bromophenyl)-5-methylbenzimidazole, 95%

Manufacturer:  Alfa Aesar™ H6403906

 View more versions of this product

Catalog No. AAH6403906

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-(4-Bromophenyl)-5-methylbenzimidazole
496807-37-5
MFCD06312161
LWVJBMHQRDXWGN-UHFFFAOYSA-N
2-(4-bromophenyl)-6-methyl-1H-benzimidazole
924735
Solid
5g
C14H11BrN2
2-4-bromophenyl-5-methyl-1h-benzimidazole, 2-4-bromophenyl-6-methyl-1h-benzimidazole, 2-4-bromophenyl-5-methylbenzimidazole, 2-4-bromophenyl-6-methyl-1h-1,3-benzodiazole, 2-4-bromophenyl-6-methyl-1h-benzo d imidazole, 2-4-bromophenyl-5-methyl-1h-1,3-benzodiazole, 2-4-bromophenyl-5-methyl-3h-1,3-benzodiazole
CC1=CC2=C(C=C1)N=C(N2)C3=CC=C(C=C3)Br
287.16
287.16
95%

Chemical Identifiers

496807-37-5
287.16
LWVJBMHQRDXWGN-UHFFFAOYSA-N
924735
CC1=CC2=C(C=C1)N=C(N2)C3=CC=C(C=C3)Br
C14H11BrN2
MFCD06312161
2-4-bromophenyl-5-methyl-1h-benzimidazole, 2-4-bromophenyl-6-methyl-1h-benzimidazole, 2-4-bromophenyl-5-methylbenzimidazole, 2-4-bromophenyl-6-methyl-1h-1,3-benzodiazole, 2-4-bromophenyl-6-methyl-1h-benzo d imidazole, 2-4-bromophenyl-5-methyl-1h-1,3-benzodiazole, 2-4-bromophenyl-5-methyl-3h-1,3-benzodiazole
2-(4-bromophenyl)-6-methyl-1H-benzimidazole
SDS Documents