Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Bromophenyl)-5-fluoroindole, 97%

Chemical Identifiers

Chemical Name or Material 2-(4-Bromophenyl)-5-fluoroindole
CAS 885266-74-0
Formula Weight 290.13
MDL Number MFCD05224214
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6309103

 
alfa aesar™
H6309103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6309106

 
alfa aesar™
H6309106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63091MD

 
alfa aesar™
H63091MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 290.13
Chemical Name or Material 2-(4-Bromophenyl)-5-fluoroindole
Quantity 1g
Physical Form Solid
Assay 97%
MDL Number MFCD05224214
CAS 885266-74-0
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1 2° to 8°C

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents, heat.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C