missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Bromophenyl)-5-fluoroindole, 97%

Manufacturer:  Alfa Aesar™ H6309106

 View more versions of this product

Catalog No. AAH6309106



Description

Description

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents, heat.
Specifications

Specifications

2-(4-Bromophenyl)-5-fluoroindole
885266-74-0
MFCD05224214
XAKLXLIHCVKNII-UHFFFAOYSA-N
2-(4-bromophenyl)-5-fluoro-1H-indole
24720950
Solid
5g
C14H9BrFN
2-4-bromophenyl-5-fluoroindole, 2-4-bromophenyl-5-fluoro-1h-indole
C1=CC(=CC=C1C2=CC3=C(N2)C=CC(=C3)F)Br
290.135
290.13
97%

Chemical Identifiers

885266-74-0
290.135
XAKLXLIHCVKNII-UHFFFAOYSA-N
24720950
C1=CC(=CC=C1C2=CC3=C(N2)C=CC(=C3)F)Br
C14H9BrFN
MFCD05224214
2-4-bromophenyl-5-fluoroindole, 2-4-bromophenyl-5-fluoro-1h-indole
2-(4-bromophenyl)-5-fluoro-1H-indole
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C

SDS Documents