Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Bromophenyl)-5-(1-naphthyl)-1,3,4-oxadiazole, 99%

$80.96 - $275.53

Chemical Identifiers

Chemical Name or Material 2-(4-Bromophenyl)-5-(1-naphthyl)-1,3,4-oxadiazole
CAS 68047-37-0
Molecular Formula C18H11BrN2O
Formula Weight 351.2
MDL Number MFCD00050980
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0855403

 
alfa aesar™
L0855403
1g Each for $80.96

AAL0855406

 
alfa aesar™
L0855406
5g Each for $275.53
Description & Specifications

Specifications

Formula Weight 351.2
Chemical Name or Material 2-(4-Bromophenyl)-5-(1-naphthyl)-1,3,4-oxadiazole
Assay 99%
CAS 68047-37-0
Molecular Formula C18H11BrN2O
MDL Number MFCD00050980
Melting Point 146° to 149°C
Quantity 1g