missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(4-Bromophenyl)-2-propanol, 97%, Thermo Scientific™
Manufacturer: Thermo Fisher Scientific H64919MD
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. It is a medicine intermediate.
Solubility
Slightly soluble in water (1.4 g/L) (25°C)
Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Specifications
2-(4-Bromophenyl)-2-propanol | |
1.356 g/mL | |
40°C to 45°C | |
C9H11BrO | |
1935066 | |
2-4-bromo-phenyl propan-2-ol, 2-4-bromo-phenyl-propan-2-ol, 2-4-bromophenyl propan-2-o1, 2-4-bromophenyl propan-2-ol, 2-4-bromophenyl-2-propanol, 2-4-bromophenyl-propan-2-ol, 4-bromo-alpha,alpha-dimethylbenzyl alcohol, acmc-209fdx, p-bromo-alpha,alpha-dimethylbenzyl alcohol | |
Slightly soluble in water (1.4g/L) (25°C) | |
CC(C)(O)C1=CC=C(Br)C=C1 | |
215.09 | |
215.09 | |
98% |
2077-19-2 | |
White | |
75°C to 85°C (0.1mmHg) | |
MFCD11870096 | |
1.552 | |
250mg | |
AOGYBHJTXLXRSM-UHFFFAOYSA-N | |
2-(4-bromophenyl)propan-2-ol | |
12681920 | |
Solid |
Chemical Identifiers
2077-19-2 | |
215.09 | |
AOGYBHJTXLXRSM-UHFFFAOYSA-N | |
12681920 | |
CC(C)(O)C1=CC=C(Br)C=C1 |
C9H11BrO | |
MFCD11870096 | |
2-4-bromo-phenyl propan-2-ol, 2-4-bromo-phenyl-propan-2-ol, 2-4-bromophenyl propan-2-o1, 2-4-bromophenyl propan-2-ol, 2-4-bromophenyl-2-propanol, 2-4-bromophenyl-propan-2-ol, 4-bromo-alpha,alpha-dimethylbenzyl alcohol, acmc-209fdx, p-bromo-alpha,alpha-dimethylbenzyl alcohol | |
2-(4-bromophenyl)propan-2-ol |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
EINECSNumber : 816-112-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only