missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Bromophenyl)-1-phenylbenzimidazole, 97%

$20.68 - $252.91

Chemical Identifiers

CAS 2620-76-0
Molecular Formula C19H13BrN2
Molecular Weight (g/mol) 349.231
MDL Number MFCD09261342
InChI Key DXRLALXPCIOIDK-UHFFFAOYSA-N
Synonym 2-4-bromophenyl-1-phenyl-1h-benzoimidazole, 2-4-bromo-phenyl-1-phenyl-1h-benzoimidazole, 2-4-bromophenyl-1-phenyl-1h-benzimidazole, 2-4-bromophenyl-1-phenylbenzimidazole, 2-4-bromophenyl-1-phenyl-1h-benzo d imidazole, 1h-benzimidazole, 2-4-bromophenyl-1-phenyl, 1-phenyl-2-4-bromophenyl benzimidazole, 2-4-bromophenyl-1-phenyl-1h-1,3-benzodiazole, 2-4-bromophenyl-1-phenyl-1,3-benzodiazole, zlchem 100
PubChem CID 23094070
IUPAC Name 2-(4-bromophenyl)-1-phenylbenzimidazole
SMILES C1=CC=C(C=C1)N2C3=CC=CC=C3N=C2C4=CC=C(C=C4)Br
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64880MD View Documents Alfa Aesar™
H64880MD
250mg Each for $20.68
 
AAH6488003 View Documents Alfa Aesar™
H6488003
1g Each for $60.77
 
AAH6488006 View Documents Alfa Aesar™
H6488006
5g Each for $252.91
 
Specifications

Chemical Identifiers

2620-76-0
349.231
DXRLALXPCIOIDK-UHFFFAOYSA-N
23094070
C1=CC=C(C=C1)N2C3=CC=CC=C3N=C2C4=CC=C(C=C4)Br
C19H13BrN2
MFCD09261342
2-4-bromophenyl-1-phenyl-1h-benzoimidazole, 2-4-bromo-phenyl-1-phenyl-1h-benzoimidazole, 2-4-bromophenyl-1-phenyl-1h-benzimidazole, 2-4-bromophenyl-1-phenylbenzimidazole, 2-4-bromophenyl-1-phenyl-1h-benzo d imidazole, 1h-benzimidazole, 2-4-bromophenyl-1-phenyl, 1-phenyl-2-4-bromophenyl benzimidazole, 2-4-bromophenyl-1-phenyl-1h-1,3-benzodiazole, 2-4-bromophenyl-1-phenyl-1,3-benzodiazole, zlchem 100
2-(4-bromophenyl)-1-phenylbenzimidazole

Specifications

White
250mg
C19H13BrN2
617138
DXRLALXPCIOIDK-UHFFFAOYSA-N
2-(4-bromophenyl)-1-phenylbenzimidazole
23094070
Powder
2-(4-Bromophenyl)-1-phenylbenzimidazole
162°C to 166°C
2620-76-0
MFCD09261342
2-4-bromophenyl-1-phenyl-1h-benzoimidazole, 2-4-bromo-phenyl-1-phenyl-1h-benzoimidazole, 2-4-bromophenyl-1-phenyl-1h-benzimidazole, 2-4-bromophenyl-1-phenylbenzimidazole, 2-4-bromophenyl-1-phenyl-1h-benzo d imidazole, 1h-benzimidazole, 2-4-bromophenyl-1-phenyl, 1-phenyl-2-4-bromophenyl benzimidazole, 2-4-bromophenyl-1-phenyl-1h-1,3-benzodiazole, 2-4-bromophenyl-1-phenyl-1,3-benzodiazole, zlchem 100
C1=CC=C(C=C1)N2C3=CC=CC=C3N=C2C4=CC=C(C=C4)Br
349.231
349.23
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning