missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(4-Bromophenyl)-1,3-dioxolane 98.0+%, TCI America™
Supplier: TCI America B210025G
Specifications
2-(4-Bromophenyl)-1,3-dioxolane | |
White-Yellow | |
MFCD01318950 | |
2-4-bromophenyl-1,3-dioxolane, 1-bromo-4-1,3-dioxolan-2-yl benzene, 4-bromobenzaldehyde ethylene acetal, 1,3-dioxolane,2-4-bromophenyl, 2-4'-bromophenyl-1,3-dioxolane, 1,3-dioxolane, 2-4-bromophenyl, bromophenyldioxolane, acmc-2098ib, 2-4-bromophenyl dioxolane, 4-bromobenzaldehyde ethylene ketal | |
C1COC(O1)C2=CC=C(C=C2)Br | |
229.073 | |
229.07 | |
Crystalline Lumps |
10602-01-4 | |
C9H9BrO2 | |
25 g | |
ZYIMHOWVWWHLDN-UHFFFAOYSA-N | |
2-(4-bromophenyl)-1,3-dioxolane | |
2773842 | |
≥98.0% (GC) |
Chemical Identifiers
10602-01-4 | |
229.073 | |
ZYIMHOWVWWHLDN-UHFFFAOYSA-N | |
2-(4-bromophenyl)-1,3-dioxolane |
C9H9BrO2 | |
MFCD01318950 | |
2773842 | |
C1COC(O1)C2=CC=C(C=C2)Br |
Safety and Handling
TSCA : No