missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(4-Bromophenoxy)-N,N-dimethylethylamine, ≥97%, Maybridge

$101.33 - $101.33

Chemical Identifiers

CAS 2474-07-9
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C10H14BrNO
Formula Weight 244.13
MDL Number MFCD00274471
Chemical Name or Material 2-(4-Bromophenoxy)-N, N-dimethylethylamine
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC51510DE Fisher Scientific Company
CC51510DE
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC51510DA SDS Fisher Scientific Company
CC51510DA
1g Each for $101.33
Specifications Description & Specifications

Chemical Identifiers

2474-07-9
100.0
C10H14BrNO
MFCD00274471
97%
96.5
97%
244.13
2-(4-Bromophenoxy)-N, N-dimethylethylamine

Specifications

244.13
100.0
97%
96.5
2-(4-Bromophenoxy)-N, N-dimethylethylamine
5g
C10H14BrNO
MFCD00274471
2474-07-9
97%

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.