missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(4-Bromophenoxy)-N,N-dimethylethylamine, 97%

Manufacturer:  Alfa Aesar™ H5429503

 View more versions of this product

Catalog No. AAH5429503



Description

Description

Applications
An intermediate for the synthesis of Tamoxifen.

Solubility
Soluble in chloroform, dichloromethane.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

2-(4-Bromophenoxy)-N,N-dimethylethylamine
2474-07-9
MFCD00274471
2-4-bromo-phenoxy-ethyl-dimethyl
MOVOYJFCKMYLHQ-UHFFFAOYSA-N
2-(4-bromophenoxy)-N,N-dimethylethanamine
213877
97%
1g
C10H14BrNO
UN2735
Soluble in chloroform,dichloromethane.
CN(C)CCOC1=CC=C(C=C1)Br
244.13
244.13

Chemical Identifiers

2474-07-9
244.13
MOVOYJFCKMYLHQ-UHFFFAOYSA-N
213877
CN(C)CCOC1=CC=C(C=C1)Br
C10H14BrNO
MFCD00274471
2-4-bromo-phenoxy-ethyl-dimethyl
2-(4-bromophenoxy)-N,N-dimethylethanamine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents