missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(4-Bromobutyl)-1,3-dioxolane, TCI America™

$366.67 - $1,387.25

Chemical Identifiers

CAS 87227-41-6
Molecular Formula C7H13BrO2
Molecular Weight (g/mol) 209.083
MDL Number MFCD02093454
InChI Key UQTZEBQZCBQLAN-UHFFFAOYSA-N
Synonym 5-Bromovaleraldehyde Ethylene Acetal
PubChem CID 13780200
IUPAC Name 2-(4-bromobutyl)-1,3-dioxolane
SMILES C1COC(O1)CCCCBr
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B21285G View Documents TCI America
B21285G
5g Each for $366.67
 
B212825G View Documents TCI America
B212825G
25g Each for $1,387.25
 
Specifications

Chemical Identifiers

87227-41-6
209.083
UQTZEBQZCBQLAN-UHFFFAOYSA-N
13780200
C1COC(O1)CCCCBr
C7H13BrO2
MFCD02093454
5-Bromovaleraldehyde Ethylene Acetal
2-(4-bromobutyl)-1,3-dioxolane

Specifications

Yellow
87227-41-6
MFCD02093454
UQTZEBQZCBQLAN-UHFFFAOYSA-N
2-(4-bromobutyl)-1,3-dioxolane
13780200
Clear Liquid at 20°C
2-(4-Bromobutyl)-1,3-dioxolane
5g
C7H13BrO2
5-Bromovaleraldehyde Ethylene Acetal
C1COC(O1)CCCCBr
209.083
209.08
≥95.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator