missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-[4-(Boc-amino)-1-piperidinyl]pyridine-5-boronic acid pinacol ester, 96%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific H5420303

 View more versions of this product

Catalog No. AAH5420303



Description

Description

Applications
2-[4-(Boc-amino)-1-piperidinyl]pyridine-5-boronic acid pinacol ester is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
Specifications

Specifications

2-[4-(Boc-amino)-1-piperidinyl]pyridine-5-boronic acid pinacol ester
1g
C21H34BN3O4
2-4-boc-amino-1-piperidinyl pyridine-5-boronic acid pinacol ester
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)N3CCC(CC3)NC(=O)OC(C)(C)C
403.33
403.32
132°C to 134°C
1042917-44-1
MFCD13195039
JGRVQSYVAOLRKS-UHFFFAOYSA-N
tert-butyl N-[1-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]piperidin-4-yl]carbamate
68484693
96%

Chemical Identifiers

1042917-44-1
403.33
JGRVQSYVAOLRKS-UHFFFAOYSA-N
68484693
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)N3CCC(CC3)NC(=O)OC(C)(C)C
C21H34BN3O4
MFCD13195039
2-4-boc-amino-1-piperidinyl pyridine-5-boronic acid pinacol ester
tert-butyl N-[1-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]piperidin-4-yl]carbamate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Keep cold

Documents