Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Bis(trifluoromethyl)phenylacetic acid, 97%

$92.97 - $232.04

Chemical Identifiers

Chemical Name or Material 2,4-Bis(trifluoromethyl)phenylacetic acid
CAS 177952-39-5
Molecular Formula C10H6F6O2
Formula Weight 272.15
MDL Number MFCD00042492
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2649203

  SDS
alfa aesar™
H2649203
1g Each for $232.04

AAH26492MD

  SDS
alfa aesar™
H26492MD
250mg Each for $92.97
Description & Specifications

Specifications

Formula Weight 272.15
Chemical Name or Material 2,4-Bis(trifluoromethyl)phenylacetic acid
Assay 97%
Molecular Formula C10H6F6O2
CAS 177952-39-5
Melting Point 85° to 90°C
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00042492
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.