Learn More
2,4-Bis(trifluoromethyl)benzyl bromide, 95%, Thermo Scientific™
Supplier: Thermo Fisher Scientific 436350010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2, 4-Bis(trifluoromethyl)benzyl bromide | |
94.0 | |
Colorless to Yellow | |
94% min. (GC) | |
Authentic | |
Glass Bottle | |
1-bromomethyl-2,4-bis trifluoromethyl benzene, 2,4-bis trifluoromethyl benzyl bromide, 2,4-bis trifluoromethyl benzylbromide, 2,4-bis trifluoromethyl-1-bromomethyl benzene, 2,4-bis-trifluoromethyl benzyl bromide, 2,4-bis-trifluoromethyl-benzyl bromide, 2,4-bis-trifluoromethyl-benzylbromide, acmc-1c0tm, benzene, 1-bromomethyl-2,4-bis trifluoromethyl, pubchem4879 | |
C1=CC(=C(C=C1C(F)(F)F)C(F)(F)F)CBr | |
307.03 | |
307.03 | |
95% |
140690-56-8 | |
100.0 | |
54°C | |
C9H5BrF6 | |
MFCD00010306 | |
1g | |
SWFFFUJOWAJJCH-UHFFFAOYSA-N | |
1-(bromomethyl)-2,4-bis(trifluoromethyl)benzene | |
518871 | |
Liquid |
Chemical Identifiers
140690-56-8 | |
307.03 | |
SWFFFUJOWAJJCH-UHFFFAOYSA-N | |
518871 | |
C1=CC(=C(C=C1C(F)(F)F)C(F)(F)F)CBr |
C9H5BrF6 | |
MFCD00010306 | |
1-bromomethyl-2,4-bis trifluoromethyl benzene, 2,4-bis trifluoromethyl benzyl bromide, 2,4-bis trifluoromethyl benzylbromide, 2,4-bis trifluoromethyl-1-bromomethyl benzene, 2,4-bis-trifluoromethyl benzyl bromide, 2,4-bis-trifluoromethyl-benzyl bromide, 2,4-bis-trifluoromethyl-benzylbromide, acmc-1c0tm, benzene, 1-bromomethyl-2,4-bis trifluoromethyl, pubchem4879 | |
1-(bromomethyl)-2,4-bis(trifluoromethyl)benzene |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER
GHS Signal Word: Danger
RUO – Research Use Only