missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Bis(trifluoromethyl)benzyl bromide, 95%, ACROS Organics™

Chemical Identifiers

CAS 140690-56-8
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C9H5BrF6
Formula Weight 307.03
MDL Number MFCD00010306
Synonym 1-(Bromomethyl)-2, 4-bis(trifluoromethyl)benzene
Chemical Name or Material 2, 4-Bis(trifluoromethyl)benzyl bromide
Assay ≥94.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC436350050 SDS Thermo Fisher Scientific
AC436350050
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC436350010 SDS Thermo Fisher Scientific
AC436350010
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

140690-56-8
100.0
C9H5BrF6
MFCD00010306
2, 4-Bis(trifluoromethyl)benzyl bromide
94.0
95%
307.03
1-(Bromomethyl)-2, 4-bis(trifluoromethyl)benzene
≥94.0%

Specifications

140690-56-8
100.0
Liquid
C9H5BrF6
MFCD00010306
Clear colorless to pale yellow
2, 4-Bis(trifluoromethyl)benzyl bromide
94.0
Authentic
95%
307.03
1-(Bromomethyl)-2, 4-bis(trifluoromethyl)benzene
5g
≥94.0%

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger